Tubing for rotarus® peristaltic pumps, Tygon® LMT-55
Supplier: HIRSCHMANN
Certificates
About this item
Non-DEHP and phthalate-free for laboratory appications.
- Outstanding chemical resistance
- Lot-to-Lot consistency for reproducible results
- Contains no BPA or phthalates
- Complies with FDA, NSF, 3-A, meets USP Class VI
Crystal clear and flexible, long-lasting and crack-resistant, the non-DEHP tubing delivers the same superior performance you have come to expect, but now in a formulation that contains a non-DEHP plasticizer. Handles most inorganic chemicals found in laboratory. It is non-oxidizing and non-contaminating, and less permeable than rubber tubing. The glassy-smooth inner bore helps prevent buildup to facilitate cleaning. Tubing is specially formulated for resistance to flex-fatigue and abrasion.
Temperature resistance: –50 to +74 °C
Hardness (Shore durometer A): 55
Density: 1,18