Tubing for rotarus® peristaltic pumps, PharMed® BPT
Supplier: HIRSCHMANN
Certificates
About this item
These tubes are biocompatible for sensitive pharmaceuticaland biotechnological applications with good general chemical resistance, excellent resistance to acid, alkali and oxidation, impermeable to light and UV radiation.
- Temperature resistance from -50 to 135 °C
- Hardness (Shore durometer A): 64