Thermal transfer printer labels, type B-481, StainerBondz
Supplier: Brady
Certificates
About this item
These labels coupled with R-6400 series ribbon will remain resistant when exposed to xylene and other chemicals that are commonly used in the slide staining process.
- No print removal and label buckling
- No discolouration that would cause barcodes to be unreadable
- Stays adhered to the slide