Order Entry
Finland
ContactUsLinkComponent
S-protein agarose, Novagen
S-protein agarose, Novagen
Catalog # 69704-4
CAS Number:  
S-protein agarose, Novagen
Catalog # 69704-4
CAS Number:  
Specifications
  • Particle Size:
    45 - 165 µm
  • Pk:
    10 mL
  • Description:
    S-Protein Agarose
Specifications
About this item
S-protein agarose specifically retains STag™ fusion proteins. The capacity of S-protein agarose varies and is based on the size and folding characteristics of a given target protein.
Binding can be performed in a column or batch mode. Elution is possible using slightly denaturing conditions or using a specific protease, such as thrombin or enterokinase, with appropriate recombinant proteins.