Anaerobic Systems, Jar gassing system
710-1698
- Description:Jar gassing system, 16 kg, 200×400×350 mm
- Pk:1
The Whitley jar gassing system provides perfect conditions for growing anaerobes in jars in just 2 minutes, and microaerophiles in 15 seconds.
- Full colour touch screen control panel allows real time monitoring to ensure that the criteria necessary for the creation of either anaerobic or microaerobic conditions has been met
- Easy to use so no complicated operator training is required
- Incorporates PIN code protected user access levels for additional security
The Whitley jar gassing system reduces costs when compared with using gas generating envelopes. It is perfect for the culture of small numbers of anaerobes and microaerophiles and eliminates the need for gas packs. It can be connected either to a cylinder of anaerobic mixed gas or directly to an anaerobic workstation.