Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Calcofluor white stain
Calcofluor white stain 5 mM in water fluorescent dye
Calcofluor white stain 5 mM in water fluorescent dye
Calcofluor white stain 5 mM in water fluorescent dye
Catalog # 29067.
Supplier:
Biotium
CAS Number:
Expand image
Calcofluor white stain 5 mM in water fluorescent dye
Catalog # 29067.
Supplier:
Biotium
CAS Number:
Specifications
Pk:
20 mL
Seq:
2305
Specifications
About this item
Product Details
:
:
916.98 g/mol
Storage Temperature:
Freezer
Formula:
C₄₀H₄₄N₁₂O₁₀S₂
Product Details