Order Entry
Finland
ContactUsLinkComponent
Human recombinant CIB1 (from E. coli)
Human recombinant CIB1 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant CIB1 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Store at 4 °C if entire vial will be used within 2-4 weeks. Store frozen at −20 °C for longer periods of time.For long term storage it is recommended to add 0.1% HSA or BSA. Avoid multiple freeze-thaw cycles.
  • Protein synonyms:
    Calcium and integrin-binding
  • Protein/peptide name:
    CIB1
  • Purity:
    Greater than 95.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Sequence:
    MGSSHHHHHH SSGLVPRGSH MGGSGSRLSK ELLAEYQDLT FLTKQEILLA HRRFCELLPQ EQRTVESSLR AQVPFEQILS LPELKANPFK ERICRVFSTS PAKDSLSFED FLDLLSVFSD TATPDIKSHY AFRIFDFDDD GTLNREDLSR LVNCLTGEGE DTRLSASEMK QLIDNILEES DIDRDGTINL SEFQHVISRS PDFASSFKIV L.
  • Formulation:
    The protein contains 20 mM Tris-HCl, pH-8 5mM DTT.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together