Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Rat recombinant TNF alpha (from cells)
Rat recombinant TNF alpha (from cells)
BIRBORB82023-20
:
Biorbyt
:
Rat recombinant TNF alpha (from cells)
BIRBORB82023-20
:
Biorbyt
:
Pk:
20 µG
Protein/peptide type:
Recombinant
Source:
Cells
Species:
Rat
Protein synonyms:
APC1
Protein/peptide name:
TNF alpha
Purity:
Purified.
Formulation:
Lyophilised.