Mouse recombinant mTIE-2 Fc (from CHO cells)
: Biorbyt
- Protein/peptide type:Recombinant
- Source:CHO cells
- Species:Mouse
- Storage conditions:Lyophilized sTIE-2 although stable at room temperature for weeks, should be stored desiccated below −18 °C. Upon reconstitution TIE-2 should be stored at 4 °C between 2-7 days and for future use below −18 °C. For long term storage it is recommended to add carrier protein (0. 1% HSA or BSA). Please prevent freeze/thaw cycles.
- Protein synonyms:Angiopoietin-1 receptor precursor
- Protein/peptide name:mTIE-2 Fc
- Purity:Greater than 90.0% as determined by(a)Analysis by RP-HPLC.(b)Analysis by SDS-PAGE.
- Solubility:It is recommended to reconstitute the lyophilized TIE-2 Fc Chimera in sterile water not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
- Formulation:TIE-2 Fc Chimera was lyophilized from concentrated (1 mg/ml) sterile solution containing 1x PBS.
- Tested applications:SDS-PAGE
Frequently Bought Together