Recombinant california 04/2009 (from baculovirus (insect cells))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Protein/peptide type:Recombinant
- Source:Baculovirus (Insect cells)
- Storage conditions:The Recombinant H1N1 A/California/04/2009 Recombinant should be stored at 4 °C. Do NOT Freeze!.
- Protein/peptide name:California 04/2009
- Purity:Greater than 90.0% under the conditions that would preserve its biological activity and tertiary structure.
- Formulation:The Recombinant H1N1 A/California/04/2009 solution 10mM Sodium phosphate pH-7, 150mM Sodium Chloride, and 0.005% Tween 20.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together