HIV-1 p24c Sf9 (from Baculovirus (insect cells))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Baculovirus (Insect cells)
- Storage conditions:Recombinant HIV -1 p24 °C ore sf9 although stable at 4 °C for weeks, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA). Please avoid freeze-thaw cycles.
- Protein/peptide name:HIV-1 p24c Sf9
- Purity:Greater than 90.0% as determined by HPLC analysis SDS-PAGE.
- Formulation:The protein solution contains 10mM Tris pH-8.0, 300mM NaCl and 0.01% Tween-20.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together