Order Entry
Finland
ContactUsLinkComponent
 
Human recombinant FGF12 (from E. coli)
 :  Biorbyt
 
Human recombinant FGF12 (from E. coli)
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    Aliquot and store at −20 °C or −80 °C. Avoid repeated freeze/thaw cycles.
  • Protein synonyms:
    FGF-12
  • Protein/peptide name:
    FGF12
  • Purity:
    Greater than 90% as determined by SDS-PAGE.
  • Sequence:
    MSSHHHHHH SSGLVPRGSH MESKEPQLKG IVTRLFSQQG YFLQMHPDGT IDGTKDENSD YTLFNLIPVG LRVVAIQGVK ASLYVAMNGE YLYSSDVFT PECKFKESVF ENYYVIYSST LYRQQESGRA WFLGLNKEGQ IMKGNRVKKT KPSSHFVPKP IEVCMYREQS LHEIGEKQGR RKSSGTPTM NGGKVVNQDS T.
  • Formulation:
    The FGF-12 solution (1mg/ml) contains 20 mM Tris pH-7.5, 1mM DTT, 2mM EDTA and 10% glycerol.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together