Order Entry
Finland
ContactUsLinkComponent
Human recombinant CLU (from HEK293 cells)
Human recombinant CLU (from HEK293 cells)
Supplier:  Biorbyt
Certificates
Human recombinant CLU (from HEK293 cells)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Storage conditions:
    Store lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C.The lyophilized protein remains stable until the expiry date when stored at −20 °C.
  • Protein synonyms:
    Clusterin
  • Protein/peptide name:
    CLU
  • Purity:
    Greater than 95% (SDS PAGE analyzed). Ni-NTA chromatography.
  • Solubility:
    Add 0.2 uml of deionized H2O and let the lyophilized pellet dissolve completely.
  • Sequence:
    DQTVSDNELQ EMSNQGSKYV NKEIQNAVNG VKQIKTLIEK TNEERKTLLS NLEEAKKKKE DALNETRESE TKLKELPGVC NETMMALWEE CKPCLKQTCM KFYARVCRSGS GLVGRQLEE FLNQSSPFYF WMNGDRIDSL LENDRQQTHM LDVMQDHFSRA SSIIDELFQ DRFFTREPQD TYHYLPFSLP HRRPHFFFPK SRIVRSLMPF SPYEPLNFHA MFQPFLEMIH EAQQAMDIHF HSPAFQHPPT EFIREGDDDR TVCREIRHNS TGCLRMKDQC DKCREILSVD CSTNNPSQAKLRRELDESLQ VAERLTRKYN ELLKSYQWKM LNTSSLLEQL NEQFNWVSRL ANLTQGEDQYYLRVTTVASH TSDSDVPSGV TEVVVKLFDS DPITVTVPVE VSRKNPKFME TVAEKALQEY RKKHREEGSL GSGWSHPQFE KTGHHHHHHH HGGQ.
  • Formulation:
    Filtered (0.4 umicron) and lyophilized PBS, pH 7.5.
  • Tested applications:
    SDS-PAGE , HPLC , FuncS

Specifications

Related Information

Frequently Bought Together