Order Entry
Finland
ContactUsLinkComponent
 
Recombinant Omp Pylori (from E. coli)
 :  Biorbyt
 
Recombinant Omp Pylori (from E. coli)
 :  Biorbyt

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Storage conditions:
    Two years frozen. One month in solution at room temperature.
  • Protein/peptide name:
    Omp Pylori
  • Purity:
    Greater than 95% pure as determined by 12% PAGE (Coomassie staining). Purified by proprietary chromatographic technique.
  • Formulation:
    the Omp Pylori recombinant protein is formulated in 1xPBS.
  • Tested applications:
    SDS-PAGE

 

 

Frequently Bought Together