Order Entry
Finland
ContactUsLinkComponent
Human recombinant HIF1 alpha (from E. coli)
Human recombinant HIF1 alpha (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant HIF1 alpha (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    MOP1 Recombinant Human although stable at 4 °C for 30 days, should be stored desiccated below −20 °C for periods greater than 30 days. Please avoid freeze/thaw cycles.
  • Protein synonyms:
    ARNT interacting
  • Protein/peptide name:
    HIF1 alpha
  • Purity:
    Greater than 95.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
  • Sequence:
    MEFKLELVEK LFAEDTEAKN PFSTQDTDLD LEMLAPYIPM DDDFQLRSFD QLSPLESSSA SPESASPQST VTVFQQTQIQ EPTANATTTT ATTDELKTVT KDRMEDIKIL IASPSPTHIH KETTSATSSP YRDTQSRTAS PNRAGKGVIE QTEKSHPRSP NVLSVALSQR TTVPEEELNP KILALQNAQR KRKMEHDGSL FQAVGIGTLL QQPDDHAATT SLSWKRVKGC KSSEQNGMEQ KTIILIPSDL ACRLLGQSMD ESGLPQLTSY DCEVNAPIQG SRNLLQGEEL LRALDQVN.
  • Formulation:
    The HIF1A recombinant Human solution is formulated 20 mM Tris-HCl pH-7.5 and 1mM DTT.
  • Tested applications:
    SDS-PAGE

Specifications

Related Information

Frequently Bought Together