Human HE4 (from E. coli)
Catalog # BIRBORB78514-50
Supplier: Biorbyt
CAS Number:
Specifications
- Pk:50 µG
- Source:E. coli
- Species:Human
- Storage conditions:Store at −20 °C. to −70 °C. in a manual defrost freezer.Avoid repeated freeze thaw cycles
- Protein synonyms:EDDM4
- Protein/peptide name:HE4
- Purity:> 95% by SDS-PAGE gel analysis.
- Formulation:PBS, pH 7.2, sterile filtered
- Tested applications:ELISA , WB , DOT , FuncS