Human PI3KdGST (from insect cells (Sf9))
Supplier: Biorbyt
Certificates
About this item
Specifications
- Source:Insect cells (Sf9)
- Species:Human
- Storage conditions:PI3Kd although stable at 4 °C for week, should be stored desiccated below −18 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please prevent freeze-thaw cycles.
- Biological activity:3,000 U/mg.
- Protein synonyms:Phosphoinositide 3-kinase p110d
- Protein/peptide name:PI3KdGST
- Purity:Greater than 95.0% as determined by SDS Page.
- Formulation:PI3Kd solution in 25mM Tris-HCl, pH 8.0, 50mM NaCl, 0.5mM MgCl2, and 50% glycerol.
- Tested applications:SDS-PAGE
Specifications
Frequently Bought Together