Order Entry
Finland
ContactUsLinkComponent
Human recombinant TGF beta 3 (from E. coli)
Human recombinant TGF beta 3 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant TGF beta 3 (from E. coli)
Supplier:  Biorbyt

Specifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    TGF-beta Human Recombinant although stable at 4 °C for week, should be stored at −20 °C. For long term storage it is recommended to add carrier protein (0.1% HSA or BSA).Please avoid freeze-thaw cycles.
  • Protein synonyms:
    ARVD
  • Protein/peptide name:
    TGF beta 3
  • Formulation:
    100ul purified human TGF Beta 3protein at 100ug/ml in 50mM Tris-Acetate, pH-7.5, 1mM EDTA and 20% Glycerol.
  • Tested applications:
    ELISA , WB

Specifications

Related Information

Frequently Bought Together