Order Entry
Finland
ContactUsLinkComponent
Septum adapter
Septum adapter
Catalog # HAMI31335
Supplier:  HAMILTON BONADUZ
undefined
Septum adapter
Catalog # HAMI31335
Supplier:  HAMILTON BONADUZ
Specifications
  • Description:
    Septum adapter for TLL syringe with septa
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
The septum adapter connects to any PTFE Luer lock (TLL) or Luer tip (LT) syringe. The adapter seals the syringe barrel but the sample remains accessible by piercing the replaceable septum.
Delivery: Supplied with 1 adapter and 12 septa.