Powder lances
Supplier: Sampling Systems
Certificates
About this item
The powder lance can be used to manually take large volume samples.
- Cross sectional sampling without damage
- Mostly used with cohesive powders
- Horizontal use through a sack or elsewhere vertically
Suitable for use in the pharmaceutical industries.