Corning® Culture Dishes, Nontreated, Sterile, Corning
Supplier: Corning
Certificates
About this item
Optically clear PS. For applications where cell attachment is not desired.
- With stacking beads to aid handling
- Supplied with vents to provide consistent gas exchange
- Sterilized by gamma radiation
- Certified non-pyrogenic
Specifications
- Catalog No:
- 734-1731
- 734-1728
- 734-1732
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta