NGAL (mouse) ELISA kit
Catalog # ENZOBPDKIT042
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Mouse
- Target protein:NGAL
- Description:NGAL (mouse) ELISA kit, 96 wells
- Seq:02
- Sensitivity:0,75 pg/ml (range 10 - 1000 pg/ml)
Specifications
About this item
For the quantitative determination of NGAL in mouse urine, plasma, serum, tissue extracts, or culture supernatants.
- Ready to use pre-coated plate and individual calibrators save time and reduce errors
- Fast results from up to 40 samples in duplicate in just 4 hours per 96 well plate
- Absorbance is read at 450 nm
A colorimetric immunometric ELISA kit for the rapid measurement of NGAL in mouse samples. Absorbance is read at 450nm.
Delivery: The kit contains a microwell plate coated with anti-NGAL (mouse) antibody (96 wells), 5X sample diluent, 2 containers of lyophilised mouse NGAL calibrators, 25X wash solution concentrate, biotinylated mouse NGAL antibody, HRP-streptavidin, TMB-substrate and stop solution.