Order Entry
Finland
ContactUsLinkComponent
Industrial gloves, oil repellent, ActivArmr® Hycron® 27-905
Industrial gloves, oil repellent, ActivArmr® Hycron® 27-905
Supplier:  Ansell
Certificates
undefined
Industrial gloves, oil repellent, ActivArmr® Hycron® 27-905
Supplier:  Ansell
Specifications
  • Catalog No:
  • ANSE27-905/11
  • ANSE27-905/9
  • ANSE27-905/8
  • ANSE27-905/10
  • Model:
  • 27-905
  • 27-905
  • 27-905
  • 27-905
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton/polyester
  • Cotton/polyester
  • Cotton/polyester
  • Cotton/polyester
  • Finish:
  • Fully coated
  • Fully coated
  • Fully coated
  • Fully coated
  • Cuff style:
  • Safety cuff
  • Safety cuff
  • Safety cuff
  • Safety cuff
  • EN norm:
  • EN 388:2016 (4221C)
  • EN 388:2016 (4221C)
  • EN 388:2016 (4221C)
  • EN 388:2016 (4221C)
Specifications