Storage vessel for electrodes
Catalog # 662-1155
Supplier: SI Analytics
Specifications
- Description:Measuring and storage vessel, Z 543, for pH electrodes with 12 mm diameter, with bayonet connector to avoid drying out
- Pk:1
Specifications
About this item
Plastic receptacle with squeezing ring sealing and bayonet fastener for electrodes with 12 mm diameter.