Order Entry
Finland
ContactUsLinkComponent
Angiotensin I ELISA Kits
Angiotensin I ELISA Kits
Catalog # ENZOADI900203
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Angiotensin I ELISA Kits
Catalog # ENZOADI900203
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    HRP-Streptavidin
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Species independent
  • Target protein:
    Angiotensin I
  • Description:
    Angiotensin I ELISA kit, 96 wells
  • Seq:
    02
  • Detection range:
    3,9 - 10000 pg/ml
  • Storage temperature:
    –20 °C/+4 °C
  • Sensitivity:
    4,3 pg/ml

Specifications

About this item

The Angiotensin I ELISA Kit is a colorimetric, competitive immunoassay kit for the quantitative determination of species independent Angiotensin I in plasma and serum samples.

  • Results in under 3,5 hours
  • Specific - low cross reactivity to similar peptides
  • Non radioactive - enzymatic assay format is safe and reliable
  • Easy to use liquid colour coded reagents reduce errors
  • Ready to use reagents and pre-coated plates reduce errors and save your time

Cross reactivity: Angiotensin I (100%), Angiotensin A (0,182%), Angiotensin II (0,083%), Angiotensin (1-12) (0,025%). Unaffected by Angiotensin (1-9), Angiotensin (1-7), Angiotensin III, Angiotensin IV, LVV-hemorphin 7, Bradykinin.

Delivery: Kit contains microtiter plate, conjugate, antibody, Assay Buffer 16, wash buffer concentrate, standard, TMB substrate, Stop Solution 2, and Streptavidin-HRP.