Single channel pipettes, adjustable, MLA Macro
VIST1061
- Colour code:Blue
- Volume:1 - 5 ml
- Description:MLA macro adjustable pipette, volume changes in 0,1 ml increments
- Seq:03
- Accuracy:±1,0 %
- Imprecision:<0,60 - <0,50 %
- Pk:1
This highly accurate, efficient pipette is an easy solution to large volume handling tasks such as reconstituting reagents, controls, or dilutions. The pipette allows volume changes in 0,1 ml increments within the pipette’s volume range of 1 to 5 ml.
- Factory calibrated for accurate volume delivery
- Can be easily adjusted as needed to achieve accurate performance on dense or viscous liquids
- Volume selections are clearly marked for easy identification