Orion™ Green Combination pH Electrode
662-2536
- Connection:Waterproof BNC, 1 m cable
- Ø×L:12×120 mm
- For:Dirty water
- Type:Orion™ Green GD9156BNWP, liquid-filled
- Description:pH combination electrode, Orion™ Green GD9156BNWP
- Material:Epoxy
- Diaphr.:Double junction (glass fibre)
- Electrolyte:3 M KCl (refillable)
- Pk:1
- Temp.:0...90 °C
- pH:0 - 14
This pH combination electrode is one of the first lab pH electrodes to meet RoHS requirements by containing no lead, mercury or other hazardous substances — making disposal of old electrodes easy and hassle free.
- Refillable liquid filling
- Double-junction for dirty-water testing
- Duarable epoxy body
- No disposal issues, no disposal cost
- Waterproof BNC connector fits any meter with BNC input
: Supplied with 1 m cable.