Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dow Corning® SYLGARD® 182 Silicone Elastomer
Dow Corning® SYLGARD® 182 silicone elastomer
Dow Corning® SYLGARD® 182 silicone elastomer
Dow Corning® SYLGARD® 182 silicone elastomer
Catalog # 634153X
Supplier:
GREEFF RW SILICONES LUBRICAN
CAS Number:
Certificates
Expand image
Dow Corning® SYLGARD® 182 silicone elastomer
Catalog # 634153X
Supplier:
GREEFF RW SILICONES LUBRICAN
CAS Number:
Certificates
Specifications
Pk:
5,5 kg
Seq:
4120
Specifications
About this item
Product Details
Documentation
:
Product Details
Documentation