Cleaning trolley Clino®, CR mini EM-CR
Supplier: PFENNIG REINIGUNGSTECHNIK
Certificates
About this item
The Clino® CR mini EM-CR model trolley system made of stainless steel suitable for cleanrooms and for non-sterile areas that require minimal space.
- EasyMop GMP with ergonomically optimised system box
- Highest efficiency and process reliability in single step technique
- Basic module mini with high end finish and castors (100 mm)
- EasyMop system boxes available in red, green, blue, yellow, grey and athracite
A small cleaning system with pre-preparation system EasyMop for technical cleanrooms suitable for ISO classes 7 and 8 or GMP areas C/D. An extra container for dropping of the used is mounted in the front for EM-CR2 trolley.
Delivery: Supplied with 2×system boxes EasyMop CR, 1×tray for system box EasyMop, disposal unit (9 L) and 1×stainless steel container.