Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sulphasalazine
Sulphasalazine ~95%, orange powder
Sulphasalazine ~95%, orange powder
Health
Sulphasalazine ~95%, orange powder
Supplier:
MP Biomedicals
Certificates
Expand image
Health
Sulphasalazine ~95%, orange powder
Supplier:
MP Biomedicals
About this item
CAS Number:
599-79-1
:
Formula:
C₁₈H₁₄N₄O₅S
MDL Number:
MFCD00057363
:
398.39 g/mol
Melting Pt:
260…265 °C (dec., lit.)
:
GHS08
Storage Temperature:
Ambient