Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Eosin Y
Eosin Y, orange powder C.I. 45380:2
Eosin Y, orange powder C.I. 45380:2
Eosin Y, orange powder C.I. 45380:2
Supplier:
MP Biomedicals
Certificates
Expand image
Eosin Y, orange powder C.I. 45380:2
Supplier:
MP Biomedicals
About this item
CAS Number:
15086-94-9
:
Formula:
C₂₀H₈Br₄O₅
MDL Number:
MFCD00036189
:
647.89 g/mol
Specification Test Results
Identity Test :
Passes
Specification Test Results