Diatomaceous earth, powder, acid-washed
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:61790-53-2
:7631-86-9
Formula:SiO₂
MDL Number:MFCD00132803
Melting Pt:1400 °C (lit.)
Density:13.0 lbs/ft³ (dry)
:60.08 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes