Paromomycin sulphate ≥97%, powder
Catalog # ICNA0215605625
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Form:White Powder
- Storage:Store at Room Temperature (15 - 30 °C)
- Potency:675 μg/mg
- Typical Working Concentration:100 mg/L
Specification Test Results
About this item
:
:615.6 g/mol
CAS Number:1263-89-4
Formula:C₂₃H₄₅N₅O₁₄·H₂SO₄
MDL Number:MFCD00079278