Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Manganese (II) chloride tetrahydrate
Manganese(II) chloride tetrahydrate, crystalline powder
Manganese(II) chloride tetrahydrate, crystalline powder
Manganese(II) chloride tetrahydrate, crystalline powder
Supplier:
MP Biomedicals
Certificates
Expand image
Manganese(II) chloride tetrahydrate, crystalline powder
Supplier:
MP Biomedicals
About this item
CAS Number:
13446-34-9
:
Formula:
MnCl₂·4H₂O
MDL Number:
MFCD00149792
:
197.9 g/mol
Storage Temperature:
Ambient
UN:
3077
ADR:
9,III
:
GHS07