Order Entry
Finland
ContactUsLinkComponent
Toluidine blue O, powder C.I. 52040
  ICNA0215264925
 :  
Toluidine blue O, powder C.I. 52040
  ICNA0215264925
 :  
  • Pk:
    25 g
  • Seq:
    3308
  • Identity Test :
    Passes
  • UV/VIS: λmax (H₂O):
    630 ± 4 nm
 :
 :305.83 g/mol
 :92-31-9
 :C₁₅H₁₆ClN₃S
 :MFCD00011934