Dextran sulphate sodium salt, M.W. ∼ 8,000, white powder, Reagent Grade
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9011-18-1
:
Formula:(C₆H₇Na₃O₁₄S₃)n
MDL Number:MFCD00081551
Specification Test Results
- Identity Test:Passes
- pH (1% aq soln):5.0 - 7.5
- Optical Rotation:+75 ° to 105 °
- Loss on Drying:10%
- Residue on Ignition:35 - 50%