
di-Sodium tetraborate decahydrate ~99%, white powder
Catalog # ICNA02190308.5
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:500 g
- Seq:3645
Specifications
Specification Test Results
- Identity Test:Passes
- Purity:~99%
- Moisture Content:30-40%
Specification Test Results
About this item
:
:381.37 g/mol
Boiling Pt:1575 °C (lit.)
Melting Pt:741 °C
Density:1.730 g/mL (lit.)
Storage Temperature:Ambient
EINECS:215-540-4
:GHS08
CAS Number:1303-96-4
Formula:Na₂B₄O₇·10H₂O
MDL Number:MFCD00149193