Order Entry
Finland
ContactUsLinkComponent
Hand cleansing cream, Solopol® Classic PURE
Hand cleansing cream, Solopol® Classic PURE
Catalog # 115-2345
Supplier:  Deb-STOKO
undefined
Hand cleansing cream, Solopol® Classic PURE
Catalog # 115-2345
Supplier:  Deb-STOKO
Specifications
  • Description:
    Hand cleansing cream, Solopol® Classic PURE, 2000 ml cartridge
  • Seq:
    02
  • Pk:
    4
Specifications
About this item
Traditional soap-free hand cleanser paste with scrubbing agent for heavy-duty contamination from, for example, oils, greases, carbon black, graphite, metallic dust, lubricants. For use in industrial environments.
  • Astopon® natural scrubbing agent (refined walnut shell powder) gently removes contaminants without damaging the skin
  • Contains Eucornol® skin protectant to minimise the risk of skin irritation
  • With re-fatting emoillent and Aloe vera
  • Solvent-free, colourant-free, perfume-free
Certifications: EU Ecolabel.
Ordering information: Soft bottle may only be used with Stoko® Vario® dispensers.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904