Powder Pillows, molybdenum
Catalog # 705-0158
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Mercaptoacetic acid
- Pk:1 KIT
- Description:Molybdenum HR
- Type:PP
- For unit:DR 6000, DR 5000, DR 3900 DR 3800, DR 2800, DR 2700, DR 1900
- Tests per kit:100
- Range:0,3 - 40 mg/l