Order Entry
Finland
ContactUsLinkComponent
High visibility shirt, long sleeves, Bresso
High visibility shirt, long sleeves, Bresso
Supplier:  SIOEN
Certificates
undefined
High visibility shirt, long sleeves, Bresso
Supplier:  SIOEN

Specifications

  • Catalog No:
  • 113-8605
  • 113-8604
  • 113-8603
  • 113-8602
  • 113-8609
  • 113-8608
  • 113-8607
  • 113-8606
  • 113-8612
  • 113-8611
  • 113-8610
  • 113-8601
  • Model:
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Bresso
  • Garment application:
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Industrial, Outdoor
  • Risk area:
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • HiVis
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Fabric:
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Exterior: 100% polyester
    Interior: 50% Viloft® + 50% polyester (dropneedle)
  • Weight:
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²
  • 185 g/m²

Specifications