Order Entry
Finland
ContactUsLinkComponent
VWR®, Aspergillus Glucose oxidase
VWR®, Aspergillus Glucose oxidase
Supplier:  VWR Chemicals
Certificates
undefined
VWR®, Aspergillus Glucose oxidase
Supplier:  VWR Chemicals
Certificates

Specifications

  • CAS No.:
    9001-37-0
  • Species:
    Aspergillus
  • Storage conditions:
    Freezer
  • Enzyme name:
    Glucose oxidase
  • Enzyme activity:
    100 U/mg
  • Grade:
    High purity

Specifications

Related Information