Inline cartridge check valves, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
Certificates
About this item
Inline cartridge check valves are designed to limit flow in one direction and deliver a low cracking pressure. Ideal placement for these valves would be on the effluent side of the column to prevent post-column derivatisation agents from flowing back and poisoning the column if the HPLC mobile phase pump were to shut down.
- Low cracking pressures
- Designed to limit flow to one direction
- Pressure rating up to 1000 psi (69 bar)