Capillary columns for drugs of abuse, TRACE™
Supplier: Thermo Fisher Scientific
Certificates
About this item
These columns are specifically designed for the analysis of common drugs of abuse.
The TR-DoA 5MS column is used for the analysis and determination of a range of toxicological target compounds including amphetamines, codeine and morphine.
The TR-DoA 35MS column is recommended for use in drug testing labs for the confirmation of THC.
Frequently Bought Together