Order Entry
Finland
ContactUsLinkComponent
Penicillin/Streptomycin/Glutamine solution, HyClone™
Penicillin/Streptomycin/Glutamine solution, HyClone™
Catalog # HYCLSV30082.01
CAS Number:  
undefined
Penicillin/Streptomycin/Glutamine solution, HyClone™
Catalog # HYCLSV30082.01
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Format:
    Liquid
  • Storage:
    -10° to -20° C
  • Antibiotic Spectrum:
    Broad Spectrum, active against gram negative and gram positive bacteria
  • Shelf life:
    24 months
  • Sterilization:
    0.1 µm sterile filtered

Specification Test Results

About this item

 :