Order Entry
Finland
ContactUsLinkComponent
 
Irritant
Ethyl-2-bromothiazole-4-carboxylate 96%
Catalog # ACRO454610010
CAS Number:  
undefined
Irritant
Ethyl-2-bromothiazole-4-carboxylate 96%
Catalog # ACRO454610010
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    White to light orange to light brown Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    66 to 72 °C
  • GC:
    ≥95.0 %

Specification Test Results

About this item

 :
Melting Pt:69…71 °C
 :236.08 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:100367-77-9
Formula:C₆H₆BrNO₂S
MDL Number:MFCD03788564