
3-Methoxypropionic acid 97+%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2544-06-1
:
Formula:C₄H₈O₃
MDL Number:MFCD00042654
:104.1100 g/mol
Boiling Pt:116 °C (9 mmHg)
Density:1.108 g/cm³
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together