Order Entry
Finland
ContactUsLinkComponent
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid 95%
Irritant
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid 95%
Certificates
undefined
Irritant
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid 95%

Specification Test Results

  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    150 to 156 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Related Information