
5-Pyrimidineboronic acid 97% may contain varying amounts of anhydride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:109299-78-7
:
Formula:C₄H₅BN₂O₂
MDL Number:MFCD03002366
:123.91 g/mol
Melting Pt:>300 °C
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Solid
- Infrared spectrum:Conforms
- HPLC:≥96.0 %