Kanamycin sulphate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:25389-94-0
:
Formula:C₁₈H₃₈N₄O₁₅S
MDL Number:MFCD00070253
:582.6 g/mol
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Assay:≥760 µg/mg (on dried basis)
- Loss on drying:≤4.0 %
- Solubility:Clear colorless
- TLC:Complies
- Residue after ignition:≤1.0 %
- pH:6.5 to 8.5 (1 % in water)