Order Entry
Finland
ContactUsLinkComponent
2-(Bromoacetyl)-6-methoxynaphthalene 96%
2-(Bromoacetyl)-6-methoxynaphthalene 96%
Catalog # ACRO450060050
CAS Number:  
undefined
2-(Bromoacetyl)-6-methoxynaphthalene 96%
Catalog # ACRO450060050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    110 to 114 °C
  • HPLC:
    ≥95.0 %

Specification Test Results

About this item

 :
 :279.13 g/mol
Melting Pt:112 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
 :GHS05
CAS Number:10262-65-4
Formula:C₁₃H₁₁BrO₂
MDL Number:MFCD03964189